missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELSR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115006
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CELSR2.
The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.
Specifications
| CELSR2 | |
| Polyclonal | |
| Unconjugated | |
| CELSR2 | |
| ADGRC2; adhesion G protein-coupled receptor C2; cadherin EGF LAG seven-pas G-type receptor 2; cadherin EGF LAG seven-pass G-type receptor 2; Cadherin family member 10; cadherin, EGF LAG seven-pass G-type receptor 2; cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila); CDHF10; Celsr2; EGFL2; EGF-like protein 2; EGF-like-domain, multiple 2; epidermal growth factor-like 2; epidermal growth factor-like protein 2; flamingo; Flamingo homolog; flamingo homolog 3; Flamingo1; flamingo-like protein celsr2; FLJ34118; FLJ42737; FLJ45143; FLJ45845; KIAA0279; MEGF3; mfmi1; mKIAA0279; Multiple EGF-like domains protein 3; multiple epidermal growth factor-like domains 3; multiple epidermal growth factor-like domains protein 3; seven-pass transmembrane cadherin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1952, 53883, 83465 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9HCU4, Q9QYP2, Q9R0M0 | |
| CELSR2 | |
| A synthesized peptide derived from human CELSR2(Accession Q9HCU4), corresponding to amino acid residues K2627-P2677. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction