missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELSR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
£550.00
Specifications
| Antigen | CELSR1 |
|---|---|
| Concentration | 0.05 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17149283
![]() |
Invitrogen™
PA5111365 |
100 μL |
£550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17149423
![]() |
Invitrogen™
PA5111506 |
100 μL |
£550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: PTSFRLQILN NYLQFEVSHG PSDVESVMLS GLRVTDGEWH HLLIELKNVK EDSEMKHLVT MTLDYGMDQN KADIGGMLPG LTVRSVVV Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 58%.
CELSR1 is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor like repeats and two laminin A G type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact mediated communication, with cadherin domains acting as homophilic binding regions and the EGF like domains involved in cell adhesion and receptor ligand interactions. This particular member is a developmentally regulated, neural specific gene which plays an unspecified role in early embryogenesis.Specifications
| CELSR1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| CELSR1 | |
| ADGRC1; adhesion G protein-coupled receptor C1; cadherin EGF LAG seven-pass G-type receptor 1; Cadherin family member 9; cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila); CDHF9; CELSR1; Flamingo homolog 2; FMI2; hFmi2; ME2; protocadherin flamingo 2 | |
| CELSR1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.05 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NYQ6 | |
| 9620 | |
| Recombinant Protein corresonding to Human CELSR1. Recombinant protein control fragment (Product #RP-104236). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title