Learn More
R&D Systems™ CellXVivo Human Monocyte-derived DC Differentiation Kit
Product Code. 18707707
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
18707707 1 Kit 1 set
1 options
Product Code. 18707707 Supplier R&D Systems™ Supplier No. CDK004
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Serum-Free Dendritic Cell Base Media; Recombinant Human IL-4; Recombinant Human GM-CSF; Recombinant Human TNF-alpha; Reconstitution Buffer 2
For the differentiation of monocyte-derived dendritic cells
For the differentiation of CD14+ monocytes into dendritic cells.
TRUSTED_SUSTAINABILITY
Specifications
Product Line R&D Systems
Quantity 1 Kit
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.