missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Cellulose Sani-Stick with Buffered Peptone Water
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
Description
- Cellulose Sani-Stick
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Specifications
| Product Type | Hygiene Sponge Sampling Kit |
| Media Type | Neutralizers |
| Sterility | Sterile |
| Includes | Gloves |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction