Learn More
LABPLAS Cellulose Sani-Stick with Buffered Peptone Water
Product Code. 17906649
Change view
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17906649 None Pack of 100
17956749 Gloves Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17906649 Supplier LABPLAS Supplier No. KSS67310BPW
  • ONLINE EXCLUSIVE PRICE
    £262.44
    £243.60 / Pack of 100
    Save £18.84
    7% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
  • Cellulose Sani-Stick
  • Saturated with buffered peptone water
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes None