missing translation for 'onlineSavingsMsg'
Learn More

Cellulose powder, microcrystallline, >97%, MP Biomedicals™

Product Code. 11444732 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444732 250 g 250g
11454732 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444732 Supplier MP Biomedicals Supplier No. 0219149983
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9004-34-6
Molecular Formula (C6H10O5)n

Specifications

Color White
Physical Form Powder
Assay Percent Range >97%
Quantity 250 g
Chemical Name or Material Cellulose powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.