missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm

Product Code. 15912460 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15912460 47 mm Pack of 100
15902460 25 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15912460 Supplier Sartorius Stedim Biotech™ Supplier No. 1132747N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm are recommended for a broad range of applications including liquid filtering for sample preparation.

  • Cellulose nitrate (CN), mixed cellulose ester (MCE), membrane filters are composed of cellulose nitrate and a small content of cellulose acetate.
  • Recommended for a broad range of applications such as the filtration of aqueous solutions, air/gas filtration, or sample preparation.
  • Suitable for sterile filtration, analytical sample preparation, clarification, particle analysis.
  • Cellulose nitrate filters, pore size 0.2 μm can be used for the analysis of filterability of beer (membrane filter test according to Esser, in “MEBAK – Brautechnische Analysemethoden – Würze, Bier, Mischgetränke – Methodensammlung der Mitteleuropäischen Brautechnischen Analysenkommission“ 2012, Chapter 2.20.1)
  • Sterilizable by autoclave, gamma irradiation, or ETO.
  • Non-sterile, bulk-packed.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Cellulose Nitrate
Pore Size 0.2 μm
Sterility Non-sterile
Grade 11327
Hydrophobicity Hydrophilic
No. per Pack 100/pk.
Thickness (Metric) 130 μm
Diameter (Metric) 47 mm
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.