missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cellulose microcrystalline, average particle size 50μm, Thermo Scientific Chemicals
£41.20 - £419.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10065563
|
Thermo Scientific Acros
387242500 |
250 g | Plastic Bottle |
£41.20
250g |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
10166663
|
Thermo Scientific Acros
387240010 |
1 kg | Plastic Bottle |
£104.00
1kg |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
11926991
|
Thermo Scientific Acros
387240050 |
5 kg | Plastic drum |
£419.00
5kg |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
9004-34-6 | |
MFCD00081512 |
Specifications
White | |
9004-34-6 | |
(C12H20O10)n | |
Solubility in water: insoluble. | |
1.5% max. | |
0.05% max. |
5 to 7 | |
Micro Fine Crystalline Powder | |
MFCD00081512 | |
Authentic | |
250 Micron <1% min., 75 Micron <30% min., 32 Micron >50% min. | |
Cellulose microcrystalline |
RUO – Research Use Only