missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Cellulose Dialysis Tubing Strips
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
Brand: Biodesign™ D316
Code : LC
Additional Details : Weight : 0.20000kg
Description
- Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
- Just wet in distilled water and use
- Available in 305mm strips
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Dialysis Tubing Strips | |
Cellulose | |
30.5 m | |
Transparent | |
For life sciences research |
3.5 kDa | |
28.7 mm | |
6.45 mL/cm | |
1 Each |