missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Cellulose Dialysis Tubing Roll

Product Code. 12797486
Change view
Click to view available options
MWCO:
14 kDa
3.5 kDa
8 kDa
Capacity (Metric):
0.32 mL/cm
1.91 mL/cm
19.13 mL/cm
3.57 mL/cm
6.45 mL/cm
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Capacity (Metric) Unit Size
12797486 3.5 kDa 6.45 mL/cm Each
12787466 14 kDa 1.91 mL/cm Each
12777466 14 kDa 0.32 mL/cm Each
12717476 14 kDa 19.13 mL/cm Each
12707476 14 kDa 6.45 mL/cm Each
12777476 8 kDa 0.32 mL/cm Each
12787476 8 kDa 1.91 mL/cm Each
12797476 8 kDa 3.57 mL/cm Each
12707486 8 kDa 6.45 mL/cm Each
12717486 8 kDa 19.13 mL/cm Each
12777486 3.5 kDa 1.91 mL/cm Each
12787486 3.5 kDa 3.57 mL/cm Each
12797466 14 kDa 3.57 mL/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 12797486 Supplier Biodesign™ Supplier No. D306
  • ONLINE EXCLUSIVE PRICE
    £337.37
    £248.22 / Pack of 1
    Save £89.15
    26% Off
In Stock
Add to basket
Edge
Add to basket

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 30.5m rolls
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs

Specifications

Product Type Dialysis Tubing Roll
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 15.5 mm
Length (Metric) 30.5 m
Capacity (Metric) 6.45 mL/cm
Color Transparent
Quantity 1 Each
For Use With (Application) For life sciences research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.