missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Cellulose Dialysis Tubing

Product Code. 12767476
Change view
Click to view available options
MWCO:
14 kDa
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO unitSize
12767476 14 kDa Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12767476 Supplier Biodesign™ Supplier No. D018
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies

Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 30.5m rolls or convenient 305mm strips
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14,000Da, 8,000Da, and 3,500Da molecular weight cut offs
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

Specifications

Product Type Dialysis Tubing
MWCO 14 kDa
Material Cellulose
Diameter (Metric) 49.5 mm
Capacity (Metric) 19.13 mL/cm
Color Transparent
Quantity 50 Pack
For Use With (Application) Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.