missing translation for 'onlineSavingsMsg'
Learn More

Cellulose Acetate Butyrate, Butyryl Content 35-39%

Product Code. 10127370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10127370 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127370 Supplier Thermo Scientific Acros Supplier No. 404255000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-36-8 | (C12H14O10)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Color White
CAS 9004-36-8
Physical Form Crystalline Powder
Melting Point 127.0°C to 240.0°C
Avg. Mol. Wt. or Mol. Wt. Range 70000 (Mn)
Molecular Formula (C12H14O10)n
MDL Number MFCD00081513
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ketones, organic acetates, lactates,, methylene, ethylene, propylene chlorides and, high-boiling solvents
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.475
Viscosity 17 to 28 (sec.)
Chemical Name or Material Cellulose acetate butyrate, Butyryl Content 35-39%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.