missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose acetate, average M.W. 100,000

Product Code. 10772411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10772411 25 g Plastic bottle 25g
10409500 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772411 Supplier Thermo Scientific Chemicals Supplier No. 177780250
  • ONLINE EXCLUSIVE PRICE
    £42.20
    £37.65 / 25g
    Save £4.55
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 9004-35-7 | (C6H7O5)nC2H3O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-35-7
Molecular Formula (C6H7O5)nC2H3O
MDL Number MFCD00081496

Specifications

Color White
Physical Form Powder
Merck Index 14, 1966
Quantity 25 g
Formula Weight 100,000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Chemical Name or Material Cellulose acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.