missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures

Product Code. 10156610
Change view
Click to view available options
Capacity (Metric):
1.7 mL
16 mL
3.3 mL (Well)
6.5 mL
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10156610 3.3 mL (Well) Pack of 100
10070282 16 mL Pack of 100
10345791 6.5 mL Pack of 100
10632672 1.7 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10156610 Supplier Greiner Bio-One Supplier No. 662102
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with automated systems. Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures are used for anchorage dependant cell lines.

  • Manufactured from transparent virgin polystyrene
  • Low auto fluorescence
  • Single position lids to prevent cross-contamination
  • Alphanumeric well coding
  • With hydrophobic surface for suspension cultures and hybridoma cell

Service Plan

No

Warranty

No

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 3.3 mL (Well)
Color Clear
Culture Area 1.9 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Non-treated
Type Multi-well Cell Culture Plate
Well Count 24
Includes Lid
No. per Case 100
No. per Pack 1
RNase Free Yes
Dimensions (L x W x H) 127.8 x 85.5 x16.5 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.