Learn More
Greiner Bio-One CELLSTAR 96-well, Non-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 10615591
Click to view available options
Color:
Clear
Unit Size:
Pack of 60
Product Code. 10615591
Brand: Greiner Bio-One 655185
Only null left
Add to basket
Add to basket
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
  • Individu
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 60
No. per Pack 1
Quantity 60/Cs.
Temperature Range (Metric) -20°C to +60°C
Well Design Chimney