missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellSensor™ NFKB-bla HEK293T Cell Line

Product Code. 10053253
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10053253 1 vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10053253 Supplier Invitrogen™ Supplier No. K1664

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CellSensor™ NFKB-bla HEK293T cell line

The CellSensor™ NFκB-bla HEK 293T cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor kappa B (NFκB) response element. To obtain the cell line, the NFκB-bla construct was transduced into HEK 293T cells by lentivirus. Subsequent flow cytometry was used to isolate clones responsive to Tumor Necrosis Factor alpha (TNFα). The cell line has been validated for DMSO tolerance, stimulation time, substrate loading time, and EC50 concentration of TNFα. This cell line is responsive to TNFα and can be used to probe NFκB signaling pathways, including those that are involved in apoptosis, viral defense, cancer, inflammation, and autoimmune disease.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Product Line CellSensor
Quantity 1 vial
Target Entry TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.