missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ NFKB-bla Freestyle™ HEK293F Cell Line

Product Code. 10194603
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194603 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194603 Supplier Thermo Scientific™ Supplier No. K1663

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nuclear Factor kappaB (NFκB) signaling regulates genes involved in apoptosis, viral defense, cancer, inflammation, and autoimmune disease.

Nuclear Factor kappaB (NFκB) signaling regulates genes involved in apoptosis, viral defense, cancer, inflammation, and autoimmune disease. The CellSensor™ NFκB-bla FreeStyle™ 293F Cell Line is responsive to tumor necrosis factor alpha (TNFα) and can be used to probe the NFκB signaling pathway. This cell line was validated for DMSO tolerance, incubation time with stimulant and substrate loading conditions. The CellSensor™ NFκB-bla FreeStyle™ 293F Cell Line contains a beta-lactamase reporter gene under the control of NFκB response element. The construct was transduced into FreeStyle™ 293F cells by lentivirus. This cell line is a pool isolated for response to TNFα by flow cytometry. It responds to agonist treatment as expected from literature. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Cytokines, Signaling Pathway
Product Line CellSensor
Quantity 1 Vial
Target Entry TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway
Type Dividing Cells
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.