missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ irf1-bla HEL Cell Line

Product Code. 10184503
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10184503 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10184503 Supplier Thermo Scientific™ Supplier No. K1647

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines.

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines. One of Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin-3 (IL-3), prolactin, erythropoietin (Epo), and granulocyte-macrophage colony stimulating factor (GM-CSF). JAK2 gene knock-out causes embryonic lethality due to defective erythropoiesis, suggesting the Jak2/Stat5 pathway plays important role in red blood cell formation. Recent discovery of activating mutation in JAK2 (V617F) present in high percentage of myeloproliferative disease (MPD) patients suggests Jak2/Stat5 pathway to be the potential therapeutic target for certain forms of MPD. The activated transcription factor Stat5 dimers recognize and bind to a specific palindromic DNA sequence found in the promoter region of β-casein, interferon regulatory factor-1 (irf-1) and a number of other genes.

The CellSensor™ irf1-bla HEL cell line contains a beta-lactamase reporter gene under control of the interferon regulatory factor-1 (irf1) response element stably integrated into HEL cells. HEL cells are a human erythroleukemia cell line that is growth factor independent and contains a endogenous homozygous JAK2V617F mutation. This cell line validated for IC50 and Z'-Factor under optimized conditions using Jak Inhibitor 1 (Figure 1). This cell line has also been tested under variable experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Cytokines, Signaling Pathway
Product Line CellSensor
Quantity 1 Vial
Target Entry JAK/STAT pathway, JAK2 (V617F), STAT5
Type Dividing Cells
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.