missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ irf1-bla CTLL-2 Cell Line

Product Code. 10083243
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083243 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083243 Supplier Thermo Scientific™ Supplier No. K1653

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines.

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin - 3 (IL - 3), prolactin, erythropoietin (Epo), and granulocyte - macrophage colony stimulating factor (GM - CSF). JAK2 gene knockout causes embryonic lethality due to defective erythropoiesis, suggesting that the Jak2/Stat5 pathway plays an important role in red blood cell formation. The recent discovery of an activating mutation in JAK2 (V617F) present in a high percentage of myeloproliferative disease (MPD) patients suggests that the Jak2/Stat5 pathway may be a potential therapeutic target for certain forms of MPD. The activated Stat5 transcription factor recognizes and binds to a specific palindromic DNA sequence found in the promoter region of β - casein, interferon regulatory factor - 1 (irf - 1) and a number of other genes. The CellSensor™ irf1 - bla CTLL2 Cell Line contains a beta - lactamase reporter gene under control of the irf - 1 response element stably integrated into CTLL2 cells. CTLL2 cells are a clone of cytotoxic T cells derived from a C57BL/6 mouse, and are cell - growth dependent on mouse Interleukin - 2 (mIL - 2). This cell line is validated for EC50 and Z' - factor under optimized conditions using mIL - 2. This cell line has also been tested under variable assay conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Responsiveness to pathway inhibitors was also tested. CellSensor™ irf1 - bla CTLL2 cells were stimulated with mIL - 2 in triplicate over the indicated concentration range in a 384 - well format. Cells were incubated for 5 hrs with agonist in 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2.5 hrs. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The 460/530 ratios for each replicate plotted against the indicated concentrations of mIL - 2. CellSensor™ irf1 - bla CTLL2 cells were treated with Jak Inhibitor 1 over the indicated concentration range in a 384 - well format for 30 min. Cells were then incubated for 5 hrs with mIL - 2 agonist (1 ng/ml) in 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2 hrs. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader, converted to percent inhibition relative to a set of controls (unstimulated cells and EC80 stimulated cells), and plotted against the indicated concentrations of Jak Inhibitor 1. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Cytokines, Signaling Pathway
Product Line CellSensor
Quantity 1 Vial
Target Entry JAK1, JAK/STAT (IL-2) pathway, JAK3, STAT5
Type Cell Line
Cell Line CTLL-2
System Type CellSensor™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.