Learn More
BINDER™ CELLROLL Set
Shop All BINDER Products
Click to view available options
Model:
CBF 170
CBF 260
Quantity:
4 Roller Bottles/Pk.
6 Roller Bottles/Pk.
Unit Size:
Each
Description
- Adjustable direction of rotation, speed of rotation, pause times, and interval cycles
- All drive rollers for the bottles run without vibrations on special ball bearings
- Driven by a stepper motor with a soft start and stop phase
- Control unit is outside the incubator and is connected to the interior motor unit via an electric access port
Specifications
Specifications
| Description | CELLROLL Set - modular, expandable roller bottle system for cell cultivation, 6 roller bottles |
| For Use With (Equipment) | BINDER Series CB and CBF |
| Material | Stainless Steel |
| Type | Cell Roller |
| For Use With (Application) | Cell Cultivation |
| Model | CBF 260 |
| Quantity | 6 Roller Bottles/Pk. |