Learn More
Cytiva Cellophane Sheets
Shop All Cytiva Products
Click to view available options
Length (Metric):
320 mm
Unit Size:
Pack of 50
Specifications
Specifications
| Applications | For electrophoretic transfer of Proteins from Polyacrylamide or Agarose gels to an immobilizing membrane |
| Product Type | Seal |
| Length (Metric) | 320 mm |
| Material | Cellophane |