Learn More
Nexcelom™ Cellometer X2 Fluorescent Viability Counter
Description
- Automated Fluorescent Yeast and Small Cell Counting in <60 Seconds
- The Cellometer X2 utilizes bright field imaging, fluorescent imaging and pattern-recognition software to quickly and accurately identify and count individual cells. Cell count, concentration, diameter, and % viability are automatically calculated and reported.
- The Cellometer X2 Allows Hemacytometer Users to:
- Increase throughput
- Improve consistency
- Ensure all data is correctly captured
- Count difficult cells (clumpy, irregular-shaped)
- Eliminate judgment errors, miscounts, and user-to-user variability
- Flourescent Analysis: Fluorescent dyes that stain DNA can be used to identify nucleated cells in a mixed cell population
- Use of a fluorescent dye, such as propidium iodide (PI), is highly recommended for accurate viability analysis of cell samples containing debris
- Because the Cellometer X2 can count dead cells in the red fluorescence channel, there is no interference from debris and live cells
- The Cellometer X2 Image Cytometer is specifically optimized for simple, 1-step determination of yeast concentration and viability. The Cellometer X2 is ideal for research laboratories and small and large breweries looking to automate their fermentation monitoring
- Analysis of both bright field and fluorescent images from each sample makes 1-step concentration & viability determination possible
- Automated Platelet Counting in Whole Blood
- Exclusion of Debris & Size-Based Counting
- View, Print, and Save Cell Images and Data Tables
- Dedicated On-line and On-site Applications Support
Specifications
Specifications
| For Use With (Equipment) | Many Compatible Dyes: Trypan blue, AO, PI, EB, 7AAD, AO/PI, AO/EB, Calcein AM, CFDA-AM, Calcein AM/PI, CFDA/PI |
| Scan Time | 60 seconds |