Learn More
Thermo Scientific™ CellInsight™ CX7 LED Pro High Content Screening Platform
Product Code. 17976980 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity unitSize
17976980 1 each Each
1 options
Product Code. 17976980 Supplier Thermo Scientific™ Supplier No. HCSDCX7LEDPRO
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Thermo Scientific CellInsight CX7 Pro High Content Screening (HCS) Platform is a fast, LED engine-based, automated cellular imaging and analysis platform designed for life scientists whom require on-the-fly quantitative analysis.
  • A fast, LED engine-based, automated cellular imaging and analysis platform designed for life scientists whom require on-the-fly quantitative analysis.
  • The CellInsight CX7 Pro HCS Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes
Features of the CellInsight CX7 Pro Platform
  • Back-illuminated sCMOS camera offer a 95% peak quantum efficiency and -20°C cooling
  • New validated applications including Cell Painting and FirePlex-HT immunoassay
  • Olympus X-line objective imaging to improve imaging quality and assay performance
  • EurekaScan Finder double-pass functionality for on-the-fly seek and find of spheroids, tissue, and rare cellular events
  • HCS Studio 5.0 with over 40 life science bio-applications and real-time analysis for fastest time-to-data possible
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning
Capabilities
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio 5.0 Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler (not included)
TRUSTED_SUSTAINABILITY
Specifications
Product Line CellInsight
Shaking Linear, Orbital, Double Orbital
Camera Highly Sensitive Monochrome CMOS
Quantity 1 each
Voltage 0 to 3000 VDC, 0 to 250 VAC