Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™
BIOPHARMA
Product Code. 15383581
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
6 x 1 L
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15383581 6 x 100 mL Pack of 6
15393581 6 x 500 mL Pack of 6
15303711 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15383581 Supplier Corning Media Tech Supplier No. 46000CI
Wastewater Testing Savings
Use promo code 28023
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Used in RNase, DNase and Protease free applications. Corning™ Molecular Biology Grade Water Tested to USP Sterile water is used for Buffer preparation, DNA/RNA purification, DNA sequencing and libraries, recombinant DNA, cloning, gene mapping.
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 6 x 100 mL