missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Celleste™ GPU Acceleration Module for 3D Deconvolution, network license (includes one seat)

Product Code. 16618987 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 controller
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16618987 1 controller Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16618987 Supplier Invitrogen™ Supplier No. AMEP4919
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste GPU Acceleration Module for Network License enables 3D deconvolution and analysis in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC.

The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste GPU Acceleration Module for Network License enables 3D deconvolution and analysis in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC. or workstation. It also helps enable the processing of thousands of voxels at once for faster processing speed, increased performance, and better response times. Coupled with the powerful image acquisition capabilities of the EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data. A Network Server License is required for use with this module.

Celleste 5.0 Image Analysis Software Network License [1] is a full-feature image analysis package designed for a range of biological applications, from image adjustments and processing with manual and automatic measurements over multiple channels, to segmentation and classification tools that help you transform images into quantitative data in a streamlined and customizable workflow. The Celleste 5.0 Image Analysis Network Server License is designed for environments where the use of multiple copies is desired. The Network Server License comes with one seat and additional Soft License Seats [2] can be added at any time. Optional modules for 2D and 3D deconvolution, visualization, analysis and GPU-powered processing allow you to customize the capabilities according to your needs. The Celleste GPU Acceleration Module for Network License helps perform 3D deconvolution and analysis in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC or workstation. It also helps enable the processing of thousands of voxels at once for faster processing speed, increased performance, and better response times.Coupled with the powerful image acquisition capabilities of the EVOS microscopes [3], Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data.Note that Celleste 5.0 Image Analysis Network Server License is required for use with any of the modules.Celleste 5.0 software with the GPU Acceleration Module offers you these important advantages:• Streamlined workflow tools that make it easy to visualize and edit images• Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images• GPU-accelerated deconvolution to increase performance and reduce the time to results• Clear images in a fraction of the time with CPU-based processingMore about the deconvolution modulesDeconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.[1] Celleste 5.0 Image Analysis Network Server License [AMEP4868][2] Celleste Image Analysis Network Seat [AMEP4869]Explore the entire EVOS line of imaging systems, accessories, and offers [3] [www.thermofisher.com/EVOS]For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Celleste
For Use With (Equipment) EVOS M5000, EVOSM7000 and Other Imaging Systems
Content And Storage Celleste 3D Deconvolution GPU Acceleration Module Card for Network License
Quantity 1 controller
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.