missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Cell Culture Multiwell Plates
Click to view available options
Culture Area:
1.9 sq. cm
9.6 sq. cm
Capacity (Metric):
3.3 mL
16.1 mL
Unit Size:
Pack of 50
Description
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Autoclavable | Not Autoclavable |
| Bottom Shape | Flat |
| Capacity (Metric) | 3.3 mL |
| Color | Clear |
| Culture Area | 1.9 sq. cm |
| Material | Polystyrene |
| Shape | Rectangular |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction