missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Poly-D-lysine Coated Cell Culture Dish
Click to view available options
Diameter (Metric):
60 mm
100 mm
Unit Size:
Pack of 100
Pack of 40
Description
Greiner Bio-One CELLCOAT Poly-D-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, as well as serum-free or -reduced cell culture.
- Source: Synthetic
- Cultivation of neuronal cells, transfected cells and other fastidious cell lines
- Cultivation in serum-free and serumreduced medium
- Improvement of cell attachment
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 28 mL |
| Color | Clear |
| Culture Area | 21 sq. cm |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Non-sterile |
| Show More |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction