missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CellAdhere™ Type I Collagen, Bovine, Solution

Product Code. 17159231
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159231 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17159231 Supplier STEMCELL Technologies Supplier No. 07001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified bovine collagen for tissue engineering research, cell culture, and biochemistry

  • Type I Collagen is the most abundant type of collagen in the human body and is a major structural protein prevalent in skin, bone, tendon, and other fibrous connective tissues
  • All collagens are characterized by a secondary protein structure consisting of three polypeptide chains wrapped around one another in a triple helical arrangement
  • CellAdhere™ Type I Collagen, Bovine, Solution is prepared from collagen extracted from bovine hide and contains a high monomer content
  • It is supplied at a concentration of approximately 6 mg/mL (0.6%) aqueous solution in 0.01 M HCl (pH at about 2.0)
  • Starting material was isolated from a closed herd and purified using a highly controlled manufacturing process
  • This process contains validated steps to ensure inactivation of possible prion and/or viral contaminants
  • Product is sterility tested
  • Endotoxin level is 0.1 EU/mL or less, as assayed by Limulus Amoebocyte Lysate assay

Specifications

Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.