missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Cell Therapy Systems Hibernate™-E CTS™ Medium

Description
Cell Therapy Systems Hibernate™-E CTS™ media is a serum-free, nutrient basal media for the short-term maintenance of embryonic tissue in ambient CO2 conditions (Ref 1). Human tissues maintained in Hibernate™-E media include brain tumors (Ref 2), umbilical cords, and foreskins.
Quick and Easy Shipment of Human Tissues
Cell Therapy Systems Hibernate™-E CTS™ media can be used for shipment of human embryonic tissue as brain tumor, umbilical cord, and foreskin at ambient temperature using a simple 3 step process:
Collect tissue in a conical tube containing Hibernate medium (brain tumor cells require additional supplements).
Ship tissues in Hibernate™ media within 24 to 72 hours on cold packs.
Transfer received tissues into media of choice and initiate cultivation.
Simplify Your Transition into Clinical Research
The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ Cell Therapy Systems (CTS™) products deliver:
Reduced burden in qualifying reagents
Included appropriate certificates of analyses and origin
Convenient access to our Drug Master File (DMF)
References:
Brewer, G.J. and Price, P.J. (1996).Viable cultured neurons in ambient carbon dioxide and hibernation storage for a month. Neuroreports 7: 1509-1512.
Brewer, G.J. and LeRoux, P.D.(2007). Human primary brain tumor cell growth inhibition in serum-free medium optimized for neuron survival. Brain Res. 1157:156-66.
Quick and Easy Shipment of Human Tissues
Cell Therapy Systems Hibernate™-E CTS™ media can be used for shipment of human embryonic tissue as brain tumor, umbilical cord, and foreskin at ambient temperature using a simple 3 step process:
Collect tissue in a conical tube containing Hibernate medium (brain tumor cells require additional supplements).
Simplify Your Transition into Clinical Research
The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ Cell Therapy Systems (CTS™) products deliver:
References:
Specifications
Specifications
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Form | Liquid |
| Product Type | CTS Medium |
| Serum Level | Serum-free |
| With Additives | Phenol Red |
| Product Line | CTS, Hibernate |
| Quantity | 500 mL |
| Shipping Condition | Ambient |
| Tested For | Cytotoxicity, Endotoxin, Sterility, pH, Osmolality |
| Show More |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?