missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cell Therapy Systems Hibernate™-A CTS™ Medium
GREENER_CHOICE

Product Code. 12093469
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12093469 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12093469 Supplier Gibco™ Supplier No. A1370501

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cell Therapy Systems Hibernate™-A CTS™ Medium

CTS Hibernate-A Medium is a serum-free, nutrient basal medium for the short-term maintenance of postnatal and adult human tissue in ambient CO2 conditions.

Quick and easy shipment of human tissues
CTS Hibernate-A Medium can be used for shipment of human postnatal and adult tissue such as brain tumors, umbilical cords, and foreskin at ambient temperature using a simple three step process:

1) Collect tissue in a conical tube containing CTS Hibernate-A Medium (brain tumor cells require additional supplements).
2) Ship tissues in CTS Hibernate-A Medium within 24 to 72 hours on cold packs.
3) Transfer received tissues into medium of choice and initiate cultivation.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

CTS Hibernate-A Medium can be combined with B-27 Supplements to yield a complete medium.

For other applications, we provide alternative formulations of CTS Hibernate Medium and B-27 Supplements.

For established protocols using Hibernate Medium, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type CTS Medium
Serum Level Serum-free
With Additives Phenol Red
Product Line CTS, Hibernate
Quantity 500 mL
Shipping Condition Ambient
Tested For Cytotoxicity, Endotoxin, Sterility, pH, Osmolality
Content And Storage Store in refrigerator (2–8°C) and protect from light.

Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.