missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cell-Surface Vimentin (CSV) Mouse anti-Human, Biotin, Clone:84-1, Abnova™
Biotin conjugated mouse monoclonal antibody recognizes human cell-surface vimentin (CSV).
Brand: Abnova H00007431-MB08.50ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataractSpecifications
| Cell-Surface Vimentin (CSV) | |
| Monoclonal | |
| Biotin | |
| PBS with no preservative; pH 7.4 | |
| P08670 | |
| VIM | |
| Human recombinant vimentin | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence | |
| 84-1 | |
| vimentin | |
| VIM | |
| FLJ36605 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7431 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction