Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Product Code. p-6933038
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
Product Code. 15516286
Brand: Invitrogen™ 00497593
Can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
  • Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
  • Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
For Use With (Application) Flow Cytometry, Functional Assays, Cell Culture
Product Line eBioscience
Product Type Cell Stimulation Cocktail