Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
Description
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
- A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
- Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
- Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
- Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Specifications
Specifications
| Quantity | 100 μL |
| For Use With (Application) | Flow Cytometry, Functional Assays, Cell Culture |
| Product Line | eBioscience |
| Product Type | Cell Stimulation Cocktail |