Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Product Code. 15452024
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
400µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452024 100 μL 100µL
15290035 4 x 100 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15452024 Supplier Invitrogen™ Supplier No. 00497093
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
  • Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
  • Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
Volume (Metric) 100μL
Product Line eBioscience
Product Type Cell Stimulation Cocktail
compliance-icons
Product Identifier
  • eBioscience Cell Stimulation Cocktail (500X)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol