missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Cell Line Optimization Nucleofector™ X Kit

Product Code. 11516339
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11516339 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11516339 Supplier Lonza™ Supplier No. VCO1001N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage 4°C
Vector pmaxGFP™
Efficiency Higher cell densities may cause lower Nucleofection Efficiencies.8% Efficiency, 73% Viability
For Use With (Equipment) Nucleofector™ Device
For Use With (Application) For plasmid DNA and/or siRNA
Includes Includes 0.9mL Nucleofector™ Solution L AndV, 2 x 0.2mLSupplement, 2 x 20 μg pmaxGFP Vector, 20 Certified cuvettes, 20 Plastic cuvettes, Store at 4°C, For Use With Nucleofector™ Device
No. of Reactions 20
Product Type Cell Line Optimization Nucleofector Kit
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.