missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Cell Line Nucleofector™ Kit V

Product Code. 10507935
Change view
Click to view available options
No. of Reactions:
25
Unit Size:
25 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10507935 25 25 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507935 Supplier Lonza™ Supplier No. VCA1003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for high transfection efficiency with siRNA duplexes even in suspension cells. Cell Line Nucleofector™ Kit V enables expression within hours - from transfection to analysis in a day.

  • Includes:
  • Nucleofector SolutionV
  • Supplement
  • pmax GFP Vector
  • Certified cuvettes
  • Plastic cuvettes
  • For Use With Nucleofector Device
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Vector pmaxGFP™
Efficiency Achieve transfection efficiencies of up to 90% with high cell viability
For Use With (Equipment) Nucleofector™ Device
For Use With (Application) Get up to 99% transfection efficiency with siRNA duplexes even in suspension cells, Expression within hours - from transfection to analysis in a day
No. of Reactions 25
Product Type Cell Line Nucleofector Kit V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.