missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Cell Line Nucleofector™ Kit T
Specifications
Specifications
| Efficiency | 99% Transfection efficiency with siRNA duplexes even in suspension cells. |
| For Use With (Equipment) | Nucleofector™ Device |
| Includes | Five different Cell Line Nucleofector™ Solutions: C, L, R, T, and V |
| No. of Reactions | 25 |
| Product Type | Cell Line Nucleofector Kit T |
| Quantity | 1 Ea. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?