missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Cell Culture Dishes, Tissue Culture Treated
For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Tissue Culture Treated helps in direct access to the cells.
Specifications
| Closure Color | Transparent |
|---|---|
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Color | Transparent |
Description
- Safety in handling during transportation and while working due to corrugated handling ring
- SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
- Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
- Easy differentiation of lid and dish bottom prevents unintended removal of the lid
- Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
- Available in three formats: 35, 60 and 100mm
Specifications
| Transparent | |
| Polystyrene | |
| Transparent | |
| Sterile | |
| Lid |
| Closure Included | |
| Flat | |
| Polystyrene | |
| Tissue Culture |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.