missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cell Activation Cocktail 500X
GREENER_CHOICE

Product Code. 18719066
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 18719066

Brand: Tocris Bioscience™ 5476/1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell activation cocktail

Cell Activation Cocktail 500X contains the following compounds: Monensin sodium salt (1.5 mM), Phorbol 12-myristate 13-acetate (0.0405 mM) and Ionomycin calcium salt (0.67 mM). Cocktail is provided as 1 ml of 500X concentrate in ethanol.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Target Buffers, Chelators and Reagents
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.