missing translation for 'onlineSavingsMsg'
Learn More

Celestine Blue, pure

Product Code. 10433743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10433743 10 g 10g
10424795 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10433743 Supplier Thermo Scientific Acros Supplier No. 195560100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1562-90-9 | C17H18ClN3O4 | 363.798 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1562-90-9
Molecular Formula C17H18ClN3O4
Molecular Weight (g/mol) 363.798
MDL Number MFCD00011927
InChI Key OJAMERVLRXYMPD-UHFFFAOYSA-N
PubChem CID 54684697
IUPAC Name 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride
SMILES CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-]

Specifications

Merck Index 15, 1961
Infrared Spectrum Authentic
Lambda Max 638 to 648nm
Formula Weight 363.79
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 700 min. (in water)
UV Spectrometry Complies
Color Black to Brown
Quantity 10 g
Physical Form Powder
Chemical Name or Material Celestine blue
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.