missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celecoxib, MedChemExpress
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Description
Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM.
Specifications
Specifications
| Chemical Name or Material | Celecoxib |
| CAS | 169590-42-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H14F3N3O2S |
| Quantity | 1 g |
| Synonym | SC 58635 |
| Solubility Information | DMSO : ≥ 50 mg/mL (131.11 mM) |
| SMILES | CC1=CC=C(C=C1)C2=CC(C(F)(F)F)=NN2C3=CC=C(C=C3)S(=O)(N)=O |
| Molecular Weight (g/mol) | 381.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction