missing translation for 'onlineSavingsMsg'
Learn More

Celecoxib, MedChemExpress

Product Code. 16446456
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446456 100 mg 100mg
16481409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446456 Supplier Medchem Express Supplier No. HY14398
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM.

Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM.

Specifications

Chemical Name or Material Celecoxib
CAS 169590-42-5
Color White
Purity Grade Notes Research
Molecular Formula C17H14F3N3O2S
Quantity 100 mg
Synonym SC 58635
Solubility Information DMSO : ≥ 50 mg/mL (131.11 mM)
SMILES CC1=CC=C(C=C1)C2=CC(C(F)(F)F)=NN2C3=CC=C(C=C3)S(=O)(N)=O
Molecular Weight (g/mol) 381.37
Formula Weight 381.37
Percent Purity 99.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.