missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Celecoxib

Product Code. 18772754
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772754 10 mg 10mg
18742534 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18772754 Supplier R&D Systems™ Supplier No. 3786/10
  • ONLINE EXCLUSIVE PRICE
    £171.71
    £153.30 / 10mg
    Save £18.41
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective cyclooxygenase-2 (COX-2) inhibitor

Celecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor (IC50 values are 15 and 0.04 μM for COX-1 and COX-2 respectively). Celecoxib is an anti-inflammatory with shorter plasma half-life in vivo than SC 58121. Displays chemopreventive activity in in vivo tumor models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169590-42-5
Molecular Formula C17H14F3N3O2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 381.37
Purity >99%
Molecular Weight (g/mol) 381.373
Chemical Name or Material Celecoxib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.