missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Celastrol, IL-17A (IL-17A Inhibitor)
Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Description
CAS Number 34157-83-0
Specifications
Specifications
| Formulation | Solubility:DMSO: >10 mg/mL assay:>or equal to 98% (HPLC) |
| Content And Storage | Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Buffer | Solubility:DMSO: >10 mg/mL assay: ≥ 98% (HPLC) |
| Quantity | 0.01 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction