missing translation for 'onlineSavingsMsg'
Learn More

CELA1 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18427036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427036 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427036 Supplier Novus Biologicals Supplier No. NBP178085IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CELA1 Polyclonal antibody specifically detects CELA1 in Mouse, Porcine samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELA1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias chymotrypsin-like elastase family member 1, chymotrypsin-like elastase family, member 1, EC 3.4.21, EC 3.4.21.36, ELA1elastase-1, elastase 1, pancreatic, Elastase-1
Host Species Rabbit
Immunogen CELA1 [Porcine Pancreas] (Uniprot: P00772)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1990
Target Species Mouse, Pig
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.