missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceftriaxone sodium salt, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Ceftriaxone sodium salt (Disodium ceftriaxone) is a third-generation cephalosporin antibiotic with excellent activity against many gram-negative, and reasonable activity against most gram-positive microorganisms. Anti-inflammatory and antioxidant characteristics.
Specifications
Specifications
| Chemical Name or Material | Ceftriaxone sodium salt |
| Physical Form | Solid |
| CAS | 74578-69-1 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H16N8Na2O7S3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Disodium ceftriaxone |
| Solubility Information | DMSO : 50 mg/mL (83.54 mM; Need ultrasonic) ∣H2O : ≥ 40 mg/mL (66.83 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction