missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ceftriaxone sodium salt hemiheptahydrate

Product Code. 15335008 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15335008 25 g 25g
15305008 1 g 1g
15325008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15335008 Supplier Thermo Scientific Chemicals Supplier No. 455420250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

Ceftriaxone, CAS # 104376-79-6, sodium salt hemiheptahydrate is the sodium salt of ceftriaxone, a third-generation beta-lactam antibiotic of the cephalosporin class.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Ceftriaxone is the sodium salt of ceftriaxone, a beta-lactam antibiotic of the cephalosporin class that works by inactivating penicillin-binding proteins

Applications

• Penicillin-binding protein inactivation inhibits peptidoglycan cross-linkage and leads to destruction of the cell wall and subsequent lysis

• It exhibits antibiotic activity against gram-negative bacteria

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104376-79-6
Molecular Formula C36H46N16Na4O21S6
Molecular Weight (g/mol) 1323.17
MDL Number MFCD01750405,MFCD01750405,MFCD09749665
InChI Key RGDBKADCOSIOEV-MAODNAKNSA-J
PubChem CID 131843450
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Ceftriaxone Sodium Salt Hemiheptahydrate
Melting Point 236.0°C
Infrared Spectrum Authentic
Water 10% max.
Quantity 25 g
Solubility Information Solubility in water: soluble.
IUPAC Name tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
Formula Weight 661.6
Percent Purity ≥94% (HPLC)
Chemical Name or Material Ceftriaxone sodium salt hemiheptahydrate
compliance-icons compliance-icons
Product Identifier
  • Ceftriaxone sodium salt hemiheptahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.