missing translation for 'onlineSavingsMsg'
Learn More

Melford Ceftriaxone Disodium Salt Hemi

Product Code. 30018174
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018174 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018174 Supplier Melford Supplier No. C547000.25
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ceftriaxone Disodium Salt is a third-generation cephalosporin antibiotic. The hemi (heptahydrate) form indicates the presence of water molecules in the crystal structure.

Ceftriaxone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis. This interference leads to structural instability and eventual cell lysis. Broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. May be used in microbiology research and studies focusing on antibacterial agents. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.

Specifications

Chemical Name or Material Ceftriaxone Disodium Salt Hemi (Heptahydrate)
CAS 104376-79-6
Quantity 250 mg
Molecular Formula C18H16N8Na2O7S2-3.5H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.