missing translation for 'onlineSavingsMsg'
Learn More

Ceftobiprole, MedChemExpress

Product Code. 16487367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487367 5 mg 5mg
16485486 100 mg 100mg
16443186 25 mg 25mg
16440947 50 mg 50mg
16414658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487367 Supplier Medchem Express Supplier No. HY112579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ceftobiprole (Ro 63-9141) is a broad-spectrum cephalosporin with high levels of in vitro activity against methicillin- (MRSA) and vancomycin-resistant staphylococci (VRSA) and penicillin-resistant streptococci with a MIC90 value of 2 μg/mL for MRSA. Ceftobiprole also inhibits gram-positive and gram-negative pathogens.

Ceftobiprole (Ro 63-9141) is a broad-spectrum cephalosporin with high levels of in vitro activity against methicillin- (MRSA) and vancomycin-resistant staphylococci (VRSA) and penicillin-resistant streptococci with a MIC90 value of 2 μg/mL for MRSA. Ceftobiprole also inhibits gram-positive and gram-negative pathogens.

Specifications

Chemical Name or Material Ceftobiprole
CAS 209467-52-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N8O6S2
Quantity 5 mg
Synonym Ro 63-9141 BAL 9141
Solubility Information DMSO : 5 mg/mL (9.35 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(/C=C3C(N([C@H]4CNCC4)CC/3)=O)CS[C@]2([H])[C@H](NC(/C(C5=NSC(N)=N5)=N\O)=O)C1=O)O
Molecular Weight (g/mol) 534.57
Formula Weight 534.57
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.