missing translation for 'onlineSavingsMsg'
Learn More

Ceftizoxime sodium, MedChemExpress

Product Code. 16408796
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16408796

Brand: Medchem Express HYB1596A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ceftizoxime sodium (SKF-88373) is third generation cephalosporin effective against Gram-negative and Gram-positive bacteria. It binds penicillin-binding proteins (PBPs) and inhibits the bacterial cell wall synthesis.

Ceftizoxime sodium (SKF-88373) is third generation cephalosporin effective against Gram-negative and Gram-positive bacteria. It binds penicillin-binding proteins (PBPs) and inhibits the bacterial cell wall synthesis.

Specifications

Chemical Name or Material Ceftizoxime sodium
Physical Form Solid
CAS 68401-82-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H12N5NaO5S2
For Use With (Application) COVID-19-immunoregulation
Synonym SKF-88373
Solubility Information DMSO : 15 mg/mL (37.00 mM; Need ultrasonic and warming)
SMILES O=C(C(N12)=CCS[C@]2([H])[C@H](NC(/C(C3=CSC(N)=N3)=N\OC)=O)C1=O)O[Na]
Molecular Weight (g/mol) 405.38
Formula Weight 405.38
Percent Purity 98.95%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.